Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-Glucosamine hydrochloride, 98+%

Catalog No. AC119900100
Click to view available options
Quantity:
10 g
100 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-84-2
Molecular Formula C6H13NO5·HCl

Specifications

Color Brown-Yellow to Yellow
Loss on Drying 1.0 % max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Quantity 10 g
Specific Rotation Condition + 72.50 (20.00°C c=2,H2O,5hrs.)
Specific Rotation +70.0° to +73.0° (20°C, 589nm) (c=2.5, H2O) (after 3 hrs)
Formula Weight 215.64
Percent Purity 98.0 to 102.0% (on dry substance) (Total base)
Physical Form Powder
Chemical Name or Material D-Glucosamine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.