Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Glucose, 1M aq. soln., sterile-filtered

Catalog No. AAJ60067AK
Change view
Click to view available options
Quantity:
125 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60067AK 250 mL
AAJ60067EQE 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60067AK Supplier Thermo Scientific Chemicals Supplier No. J60067AK
Add to Cart

CAS: 50-99-7 | C6H12O6 | 180.156 g/mol

D-(+)-Glucose used as a ideal in culture media component. Also used in foods and medicines and as a source of certain amino acids by fermentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Glucose used as a ideal in culture media component. Also used in foods and medicines and as a source of certain amino acids by fermentation.

Solubility
Not miscible or difficult to mix with water. Soluble in methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00148912
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Quantity 250 mL
Solubility Information Not miscible or difficult to mix with water. Soluble in methanol.
Formula Weight 180.16
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Physical Form Liquid
Chemical Name or Material D-(+)-Glucose, sterile
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.