Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-(+)-Glucose monohydrate

Catalog No. AAA110900E
Click to view available options
Quantity:
500 g
2500 g

CAS: 14431-43-7 | C6H12O6·H2O

D-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14431-43-7
Molecular Formula C6H12O6·H2O
MDL Number MFCD00149450

Specifications

Melting Point ∼83°C
Assay Percent Range 99%
Quantity 2500 g
Solubility Information Soluble in water.
Formula Weight 198.18 (180.16 Anhydrous)
Chemical Name or Material D- (+)-Glucose monohydrate
EINECSNumber 200-075-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.