Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-D-Glucose, MP Biomedicals™

Catalog No. MP218069110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218069110 10 g
MP218069125 25 g
MP218069180 100 g
MP218069191 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP218069110 Supplier MP Biomedicals, Inc Supplier No. 0218069110
Add to Cart
Add to Cart

Critical for production of proteins and lipid metabolism. β-D-Glucose, MP Biomedicals is a naturally occurring monosaccharide found in plants and serves as the primary source of energy for living organisms.

β-D-Glucose is combined into glucosides, disaccharides, oligosaccharides, polysaccharides (cellulose and starch), and glycogen. It is utilized as a metabolic intermediate by cells for either aerobic or anaerobic respiration.

D-Glucose exists in two cyclic forms, α-D-glucose and β-D-glucose, based on the position of the substituent at the anomeric center.

  • α-D-Glucose is the monomer unit in starch
  • β-D-glucose is the monomer unit in cellulose
  • When one of these anomers is added to solution, it undergoes reversible epimerization to the other via an open-chain form
  • During epimerization, the specific rotation of the solution changes gradually until it reaches equilibrium
  • The existence of multiple binding modes of this single monosaccharide has been studied as a model scaffold to design compounds with increased probability of ligand binding
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-61-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00063989
InChI Key WQZGKKKJIJFFOK-VFUOTHLCSA-N
Synonym beta-d-glucose, beta-d-glucopyranose, glucoside, b-d-glucopyranose, beta-glucose, beta-dextrose, .beta.-d-glucopyranose, b-glucose, b-d-glucose, unii-j4r00m814d
PubChem CID 64689
ChEBI CHEBI:15903
IUPAC Name (2R,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OC[C@H]1O[C@@H](O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 10 g
Formula Weight 180.16
Percent Purity ≥97%
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.