Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Glutamic acid 1-benzyl ester, 95%

Click to view available options
Quantity:
1g
5g
25g
79338-14-0
C12H15NO4
237.255
MFCD00237065
HFZKKJHBHCZXTQ-SNVBAGLBSA-N
h-d-glu-obzl,1-benzyl d-glutamate,r-4-amino-5-benzyloxy-5-oxopentanoic acid,d-glutamic acid 1-benzyl ester,7-benzyl glutamate,4r-4-amino-5-benzyloxy-5-oxopentanoic acid
7019728
(4R)-4-amino-5-oxo-5-phenylmethoxypentanoic acid
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)N
79338-14-0
C12H15NO4
237.255
MFCD00237065
HFZKKJHBHCZXTQ-SNVBAGLBSA-N
h-d-glu-obzl,1-benzyl d-glutamate,r-4-amino-5-benzyloxy-5-oxopentanoic acid,d-glutamic acid 1-benzyl ester,7-benzyl glutamate,4r-4-amino-5-benzyloxy-5-oxopentanoic acid
7019728
(4R)-4-amino-5-oxo-5-phenylmethoxypentanoic acid
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)N
Beilstein 3208599
Formula Weight 237.26
Physical Form Solid
Percent Purity 95%
Color White
Quantity 25g
Chemical Name or Material D-Glutamic acid 1-benzyl ester

storageNote1 : 2° to 8°C