Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Lactose Monohydrate (Powder/NF/EP/BP/JP), Fisher Chemical™

Catalog No. l8212 Shop All Fisherbrand Products
Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.312
MDL Number MFCD00150747
InChI Key WSVLPVUVIUVCRA-ZEAJSSQNSA-N
Synonym d-lactose monohydrate, d-+-lactosemonohydrate
PubChem CID 133126686
IUPAC Name (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O

Specifications

Melting Point 219°C
Color White
Physical Form Solid
Quantity 2.5 kg
Formula Weight 360.32
Grade BP/EP/JP/NF
Identification Pass Test (BP, EP, NF)
Loss on Drying 0.5% max. (JP/NF)
Packaging Poly Bottle
Appearance Pass Test (BP/EP/JP/NF)
Vapor Pressure Negligible
Decomposition Information >219°C
Density 1.525g/cm³
Chemical Name or Material D-Lactose Monohydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.