Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-D-(+)-Melibiose Monohydrate, MP Biomedicals™

Catalog No. 0210225625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-102-25625 25 g
ICN10225605 5 g
02-102-25610 10 g
ICN10225680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-102-25625 Supplier MP Biomedicals, Inc Supplier No. 0210225625
Add to Cart
Add to Cart

α-D-(+)-Melibiose Monohydrate used to improve the preparation of benzoylated ethyl 1-thioglycosidesto and to study amorphous disaccharides.

  • Melibiose has been used in a study to improve the preparation of benzoylated ethyl 1-thioglycosides.
  • It has also been used in a study to investigate the effect of water plasticization on the molecular mobility and crystallization tendency of amorphous disaccharides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-99-9
Molecular Formula C12H22O11·H2O
Synonym 6-O-α-D-galactopyranosyl-D-glucose, α-D-melibiose hydrate

Specifications

Color White
Quantity 25 g
Optical Rotation +135 ± 10° (c=4, water)
Formula Weight 360.3
Percent Purity 99.9%
Physical Form Powder
Chemical Name or Material α-D-(+)-Melibiose monohydrate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.