Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-D-Methylglucoside, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
97-30-3
C7H14O6
194.18
MFCD00064086
HOVAGTYPODGVJG-IECVIRLLSA-N
methyl alpha-d-glucopyranoside,methyl alpha-d-glucoside,alpha-methylglucoside,alpha-methyl-d-glucoside,alpha-methyl d-glucose ether,methyl-alpha-d-glucopyranoside,alpha-d-methylglucoside,me alpha-glc,alpha-d-methyl glucoside,unii-qcf122nf3r
7568545
(2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
COC1C(C(C(C(O1)CO)O)O)O
97-30-3
C7H14O6
194.18
MFCD00064086
HOVAGTYPODGVJG-IECVIRLLSA-N
methyl alpha-d-glucopyranoside,methyl alpha-d-glucoside,alpha-methylglucoside,alpha-methyl-d-glucoside,alpha-methyl d-glucose ether,methyl-alpha-d-glucopyranoside,alpha-d-methylglucoside,me alpha-glc,alpha-d-methyl glucoside,unii-qcf122nf3r
7568545
(2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
COC1C(C(C(C(O1)CO)O)O)O
Quantity 100g
Specific Rotation + 158.50
Loss on Drying 0.1% max. (60 to 70°C)
Color White
Boiling Point 200.0°C (0.2 mmHg)
Infrared Spectrum Authentic
Packaging Plastic bottle
Melting Point 169.0°C to 171.0°C
Specific Rotation Condition +158.50° (20°C c=10,water)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Beilstein 31, 179
Merck Index 15, 6152
Solubility Information Solubility in water: 108g/100mL (20 c). Other solubilities: practically insoluble in ether,5.2g/100g methanol (20 c),7.3g/100g alcohol 80% (17 c)
Formula Weight 194.18
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material α-D-Methylglucoside, 99%
Show More Show Less

EINECSNumber : 202-571-3

TSCA : TSCA