Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(D)-(+)-Neopterin, Tocris Bioscience™

Catalog No. p-4816788
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Stimulated by IFN-γ; marker of immune activation

Precursor of biopterin synthesis. Synthesized in response to interferon-γ stimulation; used as a marker of T helper cell-induced immune activation. Also an indicator of oxidative stress; modulates the effects of reactive oxygen species (ROS).

Chemical Identifiers

CAS 2009-64-5
Molecular Formula C9H11N5O4
Molecular Weight (g/mol) 253.218
InChI Key BMQYVXCPAOLZOK-XINAWCOVSA-N
Synonym d-+-neopterin, neopterin, d-neopterin, d-erythro-neopterin, 6-d-erythro-neopterin, l-neopterin, unii-9pld60n7so, 9pld60n7so, l-erythro-neopterin, 1s,2r-1-2-amino-4-hydroxypteridin-6-yl propane-1,2,3-triol
PubChem CID 448839
IUPAC Name 2-amino-6-[(1S,2R)-1,2,3-trihydroxypropyl]-1H-pteridin-4-one
SMILES C1=C(N=C2C(=N1)NC(=NC2=O)N)C(C(CO)O)O

Specifications

Quantity 10 mg
Formula Weight 253.21
Percent Purity >98%
Chemical Name or Material (D)-(+)-Neopterin
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.