Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
D(-)-Norgestrel, ≥99%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                100 mg
                            
                        
                            
                                1 g
                            
                        
                    Specifications
Specifications
| Packaging | Poly Bottle | 
| CAS | 797-63-7 | 
| Physical Form | Neat | 
| Linear Formula | C21H28O2 | 
| MDL Number | MFCD00199013 | 
| Synonym | 13 beta-Ethyl-17 alpha-ethynyl-17 beta-hydroxygon-4-en-3-one; 17 alpha-Ethynyl-18-homo-19-nor-testosterone; 18,19-Dinor-13 beta-ethyl-17 beta-hydroxy-4-pregnen-20-yn-3-one; 18,19-Dinor-4-pregnen-20-yn-3-one; Levonorgestrel | 
| Type | Analytical Standard | 
| Quantity | 100 mg | 
| Formula Weight | 312.45 | 
| Percent Purity | ≥99% | 
| Show More | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                