Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Norvaline 99.0+%, TCI America™
SDP

Catalog No. N067325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N067325G 25 g
N06731G 1 g
N06735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N067325G Supplier TCI America Supplier No. N067325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2013-12-9
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00008097
InChI Key SNDPXSYFESPGGJ-SCSAIBSYSA-N
Synonym d-norvaline, d--norvaline, 2r-2-aminopentanoic acid, h-d-nva-oh, d---norvaline, d-2-aminovaleric acid, d-2-aminopentanoic acid, r-2-aminovaleric acid, r-2-aminopentanoic acid, d-ape
PubChem CID 439575
ChEBI CHEBI:28804
IUPAC Name (2R)-2-aminopentanoic acid
SMILES CCC[C@@H](N)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 117.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Norvaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.