Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Pantethine, ≥90%, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
1 g
5 g
Description
Featured Industries: Vitamins, Nutraceuticals and Natural Products

Specifications
Specifications
CAS | 16816-67-4 |
Physical Form | Neat |
Linear Formula | C22H42N4O8S2 |
MDL Number | MFCD00151211 |
Synonym | Bis(N-pantothenylamidoethyl) disulfide; LBF factor; Pantethein oxidized form |
Type | Analytical Standard |
Quantity | 5 g |
Formula Weight | 554.72 |
Percent Purity | ≥90% |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction