Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(-)-Pantolactone, 99%, Thermo Scientific Chemicals

Catalog Number p-3758623
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
599-04-2
C6H10O3
130.14
MFCD00005392
SERHXTVXHNVDKA-BYPYZUCNSA-N
d---pantolactone, r-pantolactone, d-pantolactone, r-pantoyl lactone, d---pantoyl lactone, pantothenic lactone, r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, unii-j288d7o0js, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r, r---pantolactone
439368
CHEBI:16719
(3R)-3-hydroxy-4,4-dimethyloxolan-2-one
CC1(COC(=O)C1O)C
599-04-2
C6H10O3
130.14
MFCD00005392
SERHXTVXHNVDKA-BYPYZUCNSA-N
d---pantolactone, r-pantolactone, d-pantolactone, r-pantoyl lactone, d---pantoyl lactone, pantothenic lactone, r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, unii-j288d7o0js, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r, r---pantolactone
439368
CHEBI:16719
(3R)-3-hydroxy-4,4-dimethyloxolan-2-one
CC1(COC(=O)C1O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 88.0°C to 92.0°C
Color White
Boiling Point 120.0°C to 122.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 18, V,1, 22
Fieser 16,269; 17,270
Merck Index 15, 7116
Specific Rotation Condition −50.50° (25°C c=2,H2O)
Specific Rotation − 50.50
Solubility Information Solubility in water: soluble.
Formula Weight 130.14
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D(-)-Pantolactone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-963-3

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit