Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D(-)-Penicillamine, 99%, specified according to Ph.Eur. and USP

Catalog No. p-178760
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 52-67-5 | C5H11NO2S | 149.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-67-5
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00064302
InChI Key VVNCNSJFMMFHPL-VKHMYHEASA-N
Synonym d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline
PubChem CID 5852
ChEBI CHEBI:7959
IUPAC Name (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
SMILES CC(C)(C(C(=O)O)N)S

Specifications

Absorbance 0.07 max. (c=0.2, H2O, 1 cm cell) at 268nm
Melting Point 212.0°C
Color White
pH 4.5 to 5.5 (1% soln.)
Ignition Residue 0.1% max
Loss on Drying 0.5% max. (1 g, 60°C, 670 Pa) (P2O5)
Infrared Spectrum Authentic
Assay Percent Range 97.0 to 102% (USP)
Packaging Glass bottle
Linear Formula (CH3)2C(SH)CH(NH2)CO2H
Quantity 5 g
Merck Index 15, 7197
Specific Rotation Condition −65° (20°C c=5,1M NaOH,on dry)
Specific Rotation − 65.00
Solubility Information Solubility in water: 11.1% (20°C). Other solubilities: slightly soluble in ethanol,insoluble in ether,benzene,acetone,carbon tetra,chloride
Formula Weight 149.21
Percent Purity 99%
Physical Form Powder
Chemical Name or Material D(-)-Penicillamine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: If breathing is difficult,r
EINECSNumber 200-148-8
RTECSNumber YV9425000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.