Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Phenylalaninamide hydrochloride, 98%

Click to view available options
Quantity:
5g
25g
100g
71666-94-9
C9H13ClN2O
200.666
MFCD00153454
KLHLGTPNBQXSJT-DDWIOCJRSA-N
d-phenylalanine amide hydrochloride,h-d-phe-nh2 hcl,2r-2-amino-3-phenylpropanamide hydrochloride,c9h12n2o.hcl,h-d-phe-nh2.hcl,h-d-phe-nh2?cl,d-phe-nh2? currencyhcl,d-phenylalaninamide hydrochloride,d-phe-nh2? inverted exclamation markehcl,2r-2-amino-3-phenylpropionamide hydrochloride
56777394
(2R)-2-amino-3-phenylpropanamide;hydrochloride
C1=CC=C(C=C1)CC(C(=O)N)N.Cl
71666-94-9
C9H13ClN2O
200.666
MFCD00153454
KLHLGTPNBQXSJT-DDWIOCJRSA-N
d-phenylalanine amide hydrochloride,h-d-phe-nh2 hcl,2r-2-amino-3-phenylpropanamide hydrochloride,c9h12n2o.hcl,h-d-phe-nh2.hcl,h-d-phe-nh2?cl,d-phe-nh2? currencyhcl,d-phenylalaninamide hydrochloride,d-phe-nh2? inverted exclamation markehcl,2r-2-amino-3-phenylpropionamide hydrochloride
56777394
(2R)-2-amino-3-phenylpropanamide;hydrochloride
C1=CC=C(C=C1)CC(C(=O)N)N.Cl
Beilstein 3915663
Formula Weight 200.67
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material D-Phenylalaninamide hydrochloride