Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Phenylalanine methyl ester hydrochloride, 98%

Catalog No. ac442620050
Click to view available options
Quantity:
5 g
25 g

CAS: 13033-84-6 | C10H14ClNO2 | 215.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13033-84-6
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00066112
InChI Key SWVMLNPDTIFDDY-SBSPUUFOSA-N
Synonym d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
PubChem CID 11481330
IUPAC Name methyl (2R)-2-amino-3-phenylpropanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)CC1=CC=CC=C1

Specifications

Melting Point 157°C to 159°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Specific Rotation -35.76
Formula Weight 215.68
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material D-Phenylalanine methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.