Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Phenyllactic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
7326-19-4
C9H10O3
166.18
MFCD00078062
VOXXWSYKYCBWHO-MRVPVSSYSA-N
r-2-hydroxy-3-phenylpropionic acid,r-2-hydroxy-3-phenylpropanoic acid,2r-2-hydroxy-3-phenylpropanoic acid,d-+-phenyllactic acid,d-+-3-phenyllactic acid,r-phenyllactate,r-+-3-phenyllactic acid,3-phenyl-d-lactic acid,r-3-phenyllactic acid,r-,a-hydroxy-3-phenylpropionic acid
643327
CHEBI:32978
(2R)-2-hydroxy-3-phenylpropanoic acid
C1=CC=C(C=C1)CC(C(=O)O)O
7326-19-4
C9H10O3
166.18
MFCD00078062
VOXXWSYKYCBWHO-MRVPVSSYSA-N
r-2-hydroxy-3-phenylpropionic acid,r-2-hydroxy-3-phenylpropanoic acid,2r-2-hydroxy-3-phenylpropanoic acid,d-+-phenyllactic acid,d-+-3-phenyllactic acid,r-phenyllactate,r-+-3-phenyllactic acid,3-phenyl-d-lactic acid,r-3-phenyllactic acid,r-,a-hydroxy-3-phenylpropionic acid
643327
CHEBI:32978
(2R)-2-hydroxy-3-phenylpropanoic acid
C1=CC=C(C=C1)CC(C(=O)O)O
Specific Rotation + 18.5
Infrared Spectrum Authentic
Quantity 5g
Specific Rotation Condition +18.5° (20°C c=1,CH3OH)
Packaging Glass bottle
Melting Point 121°C to 125°C
Color White
Assay Percent Range 98%
Linear Formula C6H5CH2CH(OH)COOH
Formula Weight 166.18
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material D(+)-Phenyllactic acid, 98%
Show More Show Less

EINECSNumber : 230-803-3