Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-(+)-Raffinose Pentahydrate
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Specifications
Specifications
| Boiling Point | 78°C to 82°C |
| Packaging | Plastic bottle |
| Quantity | 1 kg |
| Beilstein | 4285763 |
| Formula Weight | 594.51 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | D-(+)-Raffinose Pentahydrate |
Safety and Handling
| EINECSNumber | 208-146-9 |
| RTECSNumber | LZ5851200 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction