Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-Ribose-5-phosphate barium salt hexahydrate, 99%

Catalog No. AAJ6601703
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6601703 1 g
AAJ66017MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6601703 Supplier Thermo Scientific Chemicals Supplier No. J6601703
Add to Cart

CAS: 15673-79-7 | C5H9BaO8P | 365.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15673-79-7
Molecular Formula C5H9BaO8P
Molecular Weight (g/mol) 365.42
MDL Number MFCD00047534
InChI Key VNFDCDZWBYRPMC-QHEAFLAXSA-L
PubChem CID 123134315
SMILES [Ba++].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O

Specifications

Color White to Yellow
Quantity 1 g
Beilstein 3781061
Sensitivity Hygroscopic
Optical Rotation +12° (c=1 in 0.2M HCl)
IUPAC Name barium;[(2R,3R,4R)-2,3,4-trihydroxy-5-oxopentyl] dihydrogen phosphate
Formula Weight 473.51
Percent Purity 99%
Physical Form Powder
Chemical Name or Material D-Ribose-5-phosphate barium salt hexahydrate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 239-750-0
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.