Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Sucrose, ACS reagent

Catalog No. p-3737416
Click to view available options
Quantity:
5 kg
25 kg

D(+)-Sucrose, CAS # 57-50-1, is a disaccharide used in a variety of biochemical applications such as protein purification. It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose. | CAS: 57-50-1 | C12H22O11 | 342.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • D(+)-Sucrose is a disaccharide carbohydrate
  • It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose

Applications

  • D(+)-Sucrose can be used for protein and nucleic acid purification
  • Its fatty acid esters have laboratory research applications in transdermal systems
  • It is also used for sucrose-gradient centrifugation, which may be employed for the selective isolation of Nocardia species from soil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 190.0°C to 192.0°C
Color White
Ignition Residue 0.01% max.
Loss on Drying 0.03% max. (105°C)
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 5 kg
Beilstein 31,424
Merck Index 15,9012
Specific Rotation +66.3° to +66.8° (25°C, 589nm) (c=26, H2O)
Solubility Information Solubility in water: 1970g/L (15°C). Other solubilities: moderately soluble in glycerol and pyridine
Formula Weight 342.29
Grade ACS Reagent
Insoluble Matter 0.005% max
Titratable Acid 0.0008 meq/g max
Physical Form Crystals
Chemical Name or Material D(+)-Sucrose
Show More Show Less
EINECSNumber 200-334-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.