Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol 98.0+%, TCI America™
SDP

Catalog No. A101725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A101725G 25 g
A1017500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A101725G Supplier TCI America Supplier No. A101725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 716-61-0
Molecular Formula C9H13N2O4
Molecular Weight (g/mol) 213.21
MDL Number MFCD00078126
InChI Key OCYJXSUPZMNXEN-RKDXNWHRSA-O
Synonym 1r,2r-2-amino-1-4-nitrophenyl propane-1,3-diol, chloramphenicol base, levoamine, 1r,2r---2-amino-1-4-nitrophenyl-1,3-propanediol, d---threo-2-amino-1-4-nitrophenyl-1,3-propanediol, d-threo---2-amino-1-p-nitrophenyl-1,3-propanediol, 1r,2r-2-amino-1-4-nitrophenyl-1,3-propanediol, d--threo-2-amino-1-p-nitrophenyl propane-1,3-diol, 1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1r,2r, 1,3-propanediol, 2-amino-1-p-nitrophenyl-, d-threo--
PubChem CID 69745
ChEBI CHEBI:78660
IUPAC Name (1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-aminium
SMILES [NH3+][C@H](CO)[C@H](O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 25 g
Formula Weight 212.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
EINECSNumber (9)-0956
RTECSNumber TY3100000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.