Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Trehalose, Dihydrate, Spectrum™ Chemical
SDP

Catalog No. 18602002 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail

T1097, 6138-23-4, C12H22O11·2H2O

Spectrum™ Chemical D-(+)-Trehalose, Dihydrate , is a natural alpha-linked disaccharide that is also referred to as mycose or tremalose. It is found naturally in plants and microorganisms such as grasshoppers, shrimp, butterflies and bees. It is thought to have a use in the treatment for Huntington′s and Parkinson′s disease as it possibly corrects defects in autophagy seen in these diseases.

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.33
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-DJCWUSJTNA-N
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
SMILES O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Packaging Poly Pail
Quantity 1 kg
Specific Rotation +197° to +201°
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.