Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-(+)-Trehalose, Dihydrate, Spectrum™ Chemical

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical D-(+)-Trehalose, Dihydrate , is a natural alpha-linked disaccharide that is also referred to as mycose or tremalose. It is found naturally in plants and microorganisms such as grasshoppers, shrimp, butterflies and bees. It is thought to have a use in the treatment for Huntington′s and Parkinson′s disease as it possibly corrects defects in autophagy seen in these diseases.
Specifications
Specifications
CAS Max % | 100% |
Ignition Residue | 0.05% |
Packaging | Poly Pail |
Quantity | 5 kg |
Specific Rotation | +197° to +201° |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction