Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D(+)-Tryptophan, 99%

Catalog No. AC168630250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC168630250 25 g
AC168630050 5 g
AC168631000 100 g
AC168635000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC168630250 Supplier Thermo Scientific Chemicals Supplier No. 168630250
Add to Cart

CAS: 153-94-6 | C11H12N2O2 | 204.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153-94-6
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00005647
InChI Key QIVBCDIJIAJPQS-SECBINFHSA-N
Synonym d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh
PubChem CID 9060
ChEBI CHEBI:16296
IUPAC Name (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 282.0°C to 285.0°C
Color White to Yellow
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Assay Percent Range 99%
Packaging Glass Bottle
Specific Rotation + 31.50
Solubility Information Solubility in water: 11g/L (20°C). Other solubilities: soluble in alkali hydroxides, soluble in hot alcohol, insoluble in chloroform
Specific Rotation Condition + 31.50 (24.00°C c=1,H2O)
Quantity 25 g
Formula Weight 204.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material D(+)-Tryptophan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.