Learn More
Thermo Scientific Chemicals D-Tryptophan, 99%
Description
D-Tryptophan is used for human embryonic kidney cell (HEK-293, ATCC: CRL-1573) culture. The product is a sweetener used to study the release of incretins from enteroendocrine cells triggered by sugar but not by sweeteners.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-Tryptophan is used for human embryonic kidney cell (HEK-293, ATCC: CRL-1573) culture. The product is a sweetener used to study the release of incretins from enteroendocrine cells triggered by sugar but not by sweeteners.
Solubility
Soluble in 0.5M hydrochloric acid (50 mg/ml), water (11 mg/ml at 20°C), hot alcohol, and dimethyl sulfoxide. Insoluble in chloroform.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 273°C (decomposition) |
| Beilstein | 86198 |
| Solubility Information | Soluble in 0.5M hydrochloric acid (50mg/ml),water (11mg/ml at 20°C),hot alcohol,and dimethyl sulfoxide. Insoluble in chloroform. |
| Quantity | 25 g |
| Formula Weight | 204.23 |
| Optical Rotation | +31.5° (c=1 in Water) |
| Percent Purity | 99% |
| Chemical Name or Material | D-Tryptophan |
Safety and Handling
| EINECSNumber | 205-819-9 |
| RTECSNumber | YN6129000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.