Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D2-40 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1940P Shop All Epredia Products
Change view
Click to view available options
Quantity:
0.1 mL
1 mL
7 mL
12 mL
Form:
Purified
Unpurified
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1940P 1 mL Purified
MS1940P0 0.1 mL Purified
MS1940R1 1 mL Purified
MS1940R7 7 mL Unpurified
MS1940RQ 12 mL Unpurified
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1940P Supplier Epredia Supplier No. MS1940P

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ D2-40 Mouse Monoclonal Antibody.

D2-40 is reactive to a 40kDa O-linked sialoglycoprotein in lymphatic endothelium, fetal testis, seminomas and dysgerminomas. D2-40 is useful in demonstrating lymphatic invasion by primary tumors and in vascular lesions. Publications suggest that D2-40 positivity in lymphovascular invasion was a more prognostic predictor of breast cancer than blood vessel invasion2. It is also considered useful in differential diagnosis of epithelioid malignant mesothelioma vs adenocarcinoma1.

Host Species: Mouse
Clone: D2-40
Isotype: IgG1
Species Reactivity: Human. Others not tested
Epitope: Not determined
Immunogen: O-linked sialoglycoprotein
Molecular Weight: 40kDa
Positive Control: Breast carcinomas, mesothelioma and colon carcinoma
Cellular Localization: Membranous/Cytoplasmic
Recommended for: Immunohistochemistry

Specifications

Applications Immunohistochemistry
Cellular Localization Membranous/Cytoplasmic.
Classification Monoclonal
Clone D2-40.
Conjugate Unconjugated
Formulation Antibody purified from tissue culture supernatant. Prepared in 10mM PBS, pH 7.4, with 0.2% BSA and <0.1% sodium azide..
Host Species Mouse
Immunogen O-linked sialoglycoprotein.
Molecular Weight of Antigen 40kDa.
Positive Control Breast carcinomas, mesothelioma and colon carcinoma.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store vial at 4°C. When stored at 2-8°C, this antibody is stable for 24 months..
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.