Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ D2HGDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584461 Supplier Invitrogen™ Supplier No. PA584461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTDPEALQAP NVDWLRTLRG CSKVLLRPRT SEEVSHILRH CHERNLAVNP QGGNTGMVGG SVPVFDEIIL STARMNRVLS FHSVS Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 82%.

This gene encodes D-2hydroxyglutarate dehydrogenase, a mitochondrial enzyme belonging to the FAD-binding oxidoreductase/transferase type 4 family. This enzyme, which is most active in liver and kidney but also active in heart and brain, converts D-2-hydroxyglutarate to 2-ketoglutarate. Mutations in this gene are present in D-2-hydroxyglutaric aciduria, a rare recessive neurometabolic disorder causing developmental delay, epilepsy, hypotonia, and dysmorphic features.
TRUSTED_SUSTAINABILITY

Specifications

Antigen D2HGDH
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene D2HGDH
Gene Accession No. Q8N465
Gene Alias AA408776; AA408778; AI325464; D2HGD; D2HGDH; D2hgdh {ECO:0000250; D-2-hydroxyglutarate dehydrogenase; D-2-hydroxyglutarate dehydrogenase, mitochondrial; FLJ42195; LOW QUALITY PROTEIN: D-2-hydroxyglutarate dehydrogenase, mitochondrial; MGC25181; RGD1307976; UniProtKB:Q8N465}
Gene Symbols D2HGDH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human D2HGDH. Recombinant protein control fragment (Product #RP-108447).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.