Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D4 Dopamine Receptor Monoclonal Antibody (625CT1.5.4)
GREENER_CHOICE

Catalog No. PIMA537614
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537614 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537614 Supplier Thermo Scientific Supplier No. MA537614
Add to Cart
Add to Cart

D4 Dopamine Receptor Monoclonal Antibody (625CT1.5.4)

D4 Dopamine Receptor Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen D4 Dopamine Receptor
Applications Western Blot
Classification Monoclonal
Clone 625CT1.5.4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DRD4
Gene Accession No. P21917, P51436
Gene Alias AW125663; D(2C) dopamine receptor; D(4) dopamine receptor; D4DR; D4R; D4RA; Dopamine 4 receptor; Dopamine D4 receptor; dopamine receptor 4; dopamine receptor D4; DRD4; Drd-4; seven transmembrane helix receptor
Gene Symbols DRD4
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 365-391 amino acids from the Central region of human DRD4.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13491, 1815
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.