Learn More
D4 Dopamine Receptor Polyclonal Antibody, Invitrogen™
Description
PA1-32861 detects Dopamine receptor D4 from human samples.
Specifications
Specifications
| Antigen | D4 Dopamine Receptor |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | DRD4 |
| Gene Accession No. | P21917 |
| Gene Alias | AW125663; D(2C) dopamine receptor; D(4) dopamine receptor; D4DR; D4R; D4RA; Dopamine 4 receptor; Dopamine D4 receptor; dopamine receptor 4; dopamine receptor D4; DRD4; Drd-4; seven transmembrane helix receptor |
| Gene Symbols | DRD4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.