Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ D4-GDI Monoclonal Antibody (10D774)
GREENER_CHOICE

Catalog No. 01676587
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-587 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-587 Supplier Invitrogen™ Supplier No. MA141181
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: antigen standard for ARHGDIB (transient overexpression lysate), Daudi or Jurkat.

Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen D4-GDI
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 10D774
Conjugate Unconjugated
Formulation PBS with 0.02% gelatin and 0.02% sodium azide
Gene ARHGDIB
Gene Accession No. P52566
Gene Alias ARHGDIB; D4; d4 gdi; GDI LYGDI; GDIA2; GDID4; LYGDI; Ly-GDI; RAP1GN1; Rho GDI 2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP dissociation inhibitor beta; rho GDP-dissociation inhibitor 2; Rho, GDP dissociation inhibitor (GDI) beta; rho-GDI beta; RhoGDI2
Gene Symbols ARHGDIB
Host Species Mouse
Immunogen A synthetic peptide corresponding to human D4-GDI/RhoGDI2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.