Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ D4-GDI Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109407
Description
Immunogen sequence: MTEKAPEPHV EEDDDDELDS KLNYKPPPQK SLKELQEMDK DDESLIKYKK TLLGDGPVVT DPKAPNVVVT RLTLVCESAP GPITMDLTGD LEALKKETIV LKEGSEYRVK IHFKVNRDIV SGLKYVQHTY RTGVKVDKAT FMVGSYGPRP EEYEFLTPVE EAPKGMLARG TYHNKSFFTD DDKQDHLSWE WNLSIKKEWT E.
Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
Specifications
| D4-GDI | |
| Polyclonal | |
| Unconjugated | |
| ARHGDIB | |
| ARHGDIB; D4; d4 gdi; GDI LYGDI; GDIA2; GDID4; LYGDI; Ly-GDI; RAP1GN1; Rho GDI 2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP dissociation inhibitor beta; rho GDP-dissociation inhibitor 2; Rho, GDP dissociation inhibitor (GDI) beta; rho-GDI beta; RhoGDI2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11857, 397 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52566, Q61599 | |
| ARHGDIB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-201 of human ARHGDIB (NP_001166.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction