Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ D4-GDI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119870
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119870 Supplier Invitrogen™ Supplier No. PA5119870
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Jurkat cell lysate, U937 cell lysate, HL-60 cell lysate, rat spleen tissue lysate, mouse spleen tissue lysate, human colon tissue, human tonsil tissue, SH-SY5Y. Subcellular Location: Cytosol.

Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen D4-GDI
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene ARHGDIB
Gene Accession No. P52566, Q61599
Gene Alias ARHGDIB; D4; d4 gdi; GDI LYGDI; GDIA2; GDID4; LYGDI; Ly-GDI; RAP1GN1; Rho GDI 2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP dissociation inhibitor beta; rho GDP-dissociation inhibitor 2; Rho, GDP dissociation inhibitor (GDI) beta; rho-GDI beta; RhoGDI2
Gene Symbols ARHGDIB
Host Species Rabbit
Immunogen Recombinant protein within C-terminal human D4 GDI from aa65-aa201.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11857, 362456, 397
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.