Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ D4-GDI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527873 Supplier Invitrogen™ Supplier No. PA527873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji , mouse fibroblast. Predicted reactivity: Mouse (90%), Rat (89%), Xenopus laevis (81%), Pig (94%), Chicken (92%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen D4-GDI
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene ARHGDIB
Gene Accession No. P52566, Q61599
Gene Alias ARHGDIB; D4; d4 gdi; GDI LYGDI; GDIA2; GDID4; LYGDI; Ly-GDI; RAP1GN1; Rho GDI 2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP dissociation inhibitor beta; rho GDP-dissociation inhibitor 2; Rho, GDP dissociation inhibitor (GDI) beta; rho-GDI beta; RhoGDI2
Gene Symbols ARHGDIB
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Rho GDI2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11857, 397
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.