Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D4-GDI/RhoGDI2 Antibody (10D774), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056556H
Description
D4-GDI/RhoGDI2 Monoclonal specifically detects D4-GDI/RhoGDI2 in Human samples. It is validated for Western Blot.Specifications
D4-GDI/RhoGDI2 | |
Monoclonal | |
HRP | |
ARHGDIB Rho GDP dissociation inhibitor (GDI) beta D4, GDIA2, GDID4, Ly-GDI, LYGDI, RAP1GN1, RhoGDI2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
A 28 kDa band is observed. | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
10D774 | |
Western Blot | |
ARHGDIB | |
A synthetic peptide corresponding to human D4-GDI/RhoGDI2 was used as the immunogen. | |
0.1 mL | |
Apoptosis, Signal Transduction | |
397 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction