Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D4-GDI/RhoGDI2 Antibody (10D774), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056556MFV450
Description
D4-GDI/RhoGDI2 Monoclonal antibody specifically detects D4-GDI/RhoGDI2 in Human samples. It is validated for Flow CytometrySpecifications
| D4-GDI/RhoGDI2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 10D774 | |
| Flow Cytometry | |
| ARHGDIB Rho GDP dissociation inhibitor (GDI) beta, D4, GDIA2, GDID4, Ly-GDI, LYGDI, RAP1GN1, RhoGDI2 | |
| A synthetic peptide corresponding to human D4-GDI/RhoGDI2 was used as the immunogen. | |
| 0.1 mL | |
| Apoptosis, Signal Transduction | |
| 397 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction