Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D4-GDI/RhoGDI2 Antibody (97A1015) - (cleavage specific) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 16 publications

$206.50 - $499.50

Specifications

Antigen D4-GDI/RhoGDI2
Clone 97A1015
Concentration 1.0 mg/ml
Dilution Western Blot 2 ug/ml, Flow Cytometry 0.5-1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular) 0.5-1 ug/10^6 cells
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056725T
SDP
View Documents
Novus Biologicals
NB10056725SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056725
SDP
View Documents
Novus Biologicals
NB10056725
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

D4-GDI/RhoGDI2 Monoclonal specifically detects D4-GDI/RhoGDI2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications

Specifications

D4-GDI/RhoGDI2
1.0 mg/ml
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human, Mouse
ARHGDIB Rho GDP dissociation inhibitor (GDI) beta D4, GDIA2, GDID4, Ly-GDI, LYGDI, RAP1GN1, RhoGDI2
ARHGDIB
IgG2b κ
Protein G purified
In anti-Fas treated Jurkat, an approx. 23 kDa band should be observed.
97A1015
Western Blot 2 ug/ml, Flow Cytometry 0.5-1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular) 0.5-1 ug/10^6 cells
Monoclonal
Purified
RUO
P52566
397
A synthetic peptide corresponding to the region of the Fas-induced cleavage site of human D4-GDI/RhoGDI2 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.