Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D4S234E Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208002
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8002 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8002 Supplier Fabgennix Inc Supplier No. NSG1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

D4S234E Polyclonal antibody specifically detects D4S234E in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen D4S234E
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene NSG1
Gene Accession No. P02683, P42857, Q62092
Gene Alias brain neuron cytoplasmic protein 1; brain neuron cytoplasmic protein 1/2; brain specific mRNA b; Bsmrb; carboxyterminally EE-tagged neuron-enriched endosomal 21 kDa protein; D4S234; D4S234E; m234; Neep21; neuron specific gene family member 1; Neuronal vesicle trafficking-associated protein 1; Neuron-enriched endosomal protein of 21 kDa; Neuron-specific protein family member 1; NSG1; p21; PEP1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 140-190 on human Neuron-specific protein family member 1.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18196, 25247, 27065
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.