Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D4S234E Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | D4S234E |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
D4S234E Polyclonal specifically detects D4S234E in Rat samples. It is validated for Western Blot.Specifications
D4S234E | |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS buffer, 2% sucrose | |
27065 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Rat | |
carboxyterminally EE-tagged neuron-enriched endosomal 21 kDa protein, D4S234Brain neuron cytoplasmic protein 1, DNA segment on chromosome 4 (unique) 234 expressed sequence, NEEP21, neuron-specific protein family member 1, NSG1, P21 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Rat D4S234E (NP_077042). Peptide sequence VKLGNNFAEKGTKQPLLEDGFDTIPLMTPLDVNQLQFPPPDKVVVKTKTE | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title