Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ D9Ertd402e Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570020
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%; Zebrafish: 77%.
TCAIM is a protein coding gene.
Specifications
D9Ertd402e | |
Polyclonal | |
Unconjugated | |
TCAIM | |
C22H3orf23; C3orf23; cc023; D9Ertd402e; Gm1129; RGD1560717; T cell activation inhibitor, mitochondrial; TCAIM; T-cell activation inhibitor, mitochondrial; Toag1; TOAG-1; tolerance associated gene-1 protein; tolerance-associated gene 1; Tolerance-associated gene-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
382117 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q66JZ4 | |
TCAIM | |
Synthetic peptide located within the following N-terminal region: MLGSWRAYSE MFCHLRPLRR FGLRKVLPHW LHYSRALSGA EAINALRPFY. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction