Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STERIS™ DA-7645 Detergent
Shop All STERIS Corporation Products
Click to view available options
Size:
5 gal.
55 gal.
300 gal.
Container Type:
Drum
Pail
Tote
Description
DA-7645 Process and Research Cleaner is specially formulated to enhance manual cleaning with enhanced substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.
HOW DA-7645 WORKS
DA-7645 Detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms in manual cleaning applications.
WHY DA-7645?
- Efficacy - Mild pH formulation delivers excellent cleaning characteristics and substrate compatibility with enhanced operator safety.
- Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
- Productivity - Delivers efficiency to manual cleaning operations using a blend of surfactants, solvents, and other performance-enhancing ingredients.
- Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.
Specifications
Specifications
Product Type | Detergent |
Color-coded | No |
Form | Clear Liquid |
pH | 8.9 (Diluted) |
Scent | Unscented |
Sterility | Non-sterile |
Container Type | Tote |
Size | 300 gal. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction