Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAAM1 Antibody (5D3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323272
Description
DAAM1 Monoclonal antibody specifically detects DAAM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
DAAM1 | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
5D3 | |
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:200 | |
disheveled-associated activator of morphogenesis 1, dishevelled associated activator of morphogenesis 1, dishevelled-associated activator of morphogenesis 1, KIAA0666FLJ41657 | |
Clone 5D3 was generated from a recombinant protein that included amino acid residues from the N-terminal region of human DAAM1. This sequence has less than 50% homology to the N-terminal region of DAAM2. [UniProt# Q9Y4D1] | |
0.1 mL | |
Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway | |
23002 | |
Store at -20°C. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction