Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAAM1 Monoclonal Antibody (5D3)
GREENER_CHOICE

Catalog No. PIMA547712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547712 Supplier Invitrogen™ Supplier No. MA547712
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody detects a 125 kDa* protein corresponding to the apparent molecular mass of DAAM1 on SDS-PAGE immunoblots of human A431, K562, and mouse C2C12 cells.

DAAM1 is a diaphanous-related formin first studied as a novel Disheveled binding protein and shown to be crucial for the planar cell polarity pathway in Xenopus. DAAM1, like other formins, directs nucleation and elongation of new actin filaments using its conserved formin-homology-2 (FH2) domain. It is a Rho-regulated formin implicated in neuronal development and regulation of gastrulation. It was identified as a binding protein of the Wnt receptor-associated protein Disheveled. It also binds the GTP-bound RhoA and increases its GTP-bound form population in cells. It exists in an autoinhibited state via intramolecular interactions between its amino-terminal GBD and carboxyl-terminal DAD domains. Structurally the protein includes a GTPase-binding domain (GBD), a diaphanous inhibitory domain (DID), and a coiled-coil region followed by the FH1, FH2 and C-terminal diaphanous auto-regulatory domain (DAD). In humans, the DAAM1 gene is mapped to chromosome 14q23.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAAM1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5D3
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene DAAM1
Gene Accession No. Q8BPM0, Q9Y4D1
Gene Alias Daam1; Disheveled-associated activator of morphogenesis 1; dishevelled associated activator of morphogenesis 1; I79_021650; KIAA0666
Gene Symbols DAAM1
Host Species Mouse
Immunogen Clone 5D3 was generated from a recombinant protein that included amino acid residues from the N-terminal region of human DAAM1. This sequence has less than 50% homology to the N-terminal region of DAAM2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208846, 23002, 314212
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.