Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAB EASY tablets, for immunohistochemistry, 10 mg/tablet

Catalog No. ac328005000
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

DAB, or 3,3-diaminobenzidine tetrahydrochloride, EASY tablets for immunohistochemistry, CAS # 7411-49-6, is a chromogenic substrate used to observe peroxidase activity. In immunohistochemistry, DAB is used to catalyze peroxidase-bound antibodies in order to visualize protein presence. | CAS: 7411-49-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 3,3′-Diaminobenzidine is a derivative of benzene and the precursor to polybenzimidazole

Applications

  • DAB is mainly used to visualize peroxidase activity. During this chemical reaction, DAB is a hydrogen donor in the presence of peroxide. When DAB is oxidized, an insoluble brown end-product is produced.
  • DAB has been used for staining and oxidative detection of mitochondria
  • It has also been utilized for electron microscopy applications
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Tablets
Chemical Name or Material DAB EASY tablets
CAS 7411-49-6, 10043-35-3
Packaging Plastic bottle
Solubility Solubility in water: soluble.
Quantity 500 mg
MDL Number MFCD00007725

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.