Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dab1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981464
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1464 Supplier Bioss Supplier No. BS11547R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The laminar organization of multiple neuronal types in the cerebral cortex is required for normal cognitive function. In mice, the disabled-1 gene plays a central role in brain development, directing the migration of cortical neurons past previously formed neurons to reach their proper layer. This gene is similar to disabled-1, and the protein encoded by this gene is thought to be a signal transducer that interacts with protein kinase pathways to regulate neuronal positioning in the developing brain. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.

Specifications

Antigen Dab1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DAB1
Gene Accession No. O75553
Gene Alias AI956902; C630028C02Rik; Dab reelin signal transducer 1; Dab, reelin signal transducer, homolog 1; Dab, reelin signal transducer, homolog 1 (Drosophila); DAB1; DAB1, reelin adaptor protein; disabled 1; Disabled homolog 1; RP23-356E21.1; scm; scr; scrambler; yot; yotari
Gene Symbols DAB1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Dab1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1600
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.