Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dab1, Rat anti-Mouse, Clone: 4E12, Millipore Sigma™
Rat Monoclonal Antibody
$192.34 - $482.80
Specifications
Antigen | Dab1 |
---|---|
Clone | 4E12 |
Dilution | Western Blotting Analysis: A representative lot detected Dab1 in Western Blotting applications (Onoune, A., et. al. (2014). Neurosci Res. 88:23-27; Nakamura, K., et. al. (2016). Neuroscience. 336:20-29).Immunoprecipitation Analysis: A 1:500 dilution from a representative lot immunoprecipitated Dab1 in cerebral cortex tissue lysate from P20 mouse (Courtesy of Prof. Mitsuharu Hattori, Nagoya City University).Immunohistochemistry Analysis: A representative lot detected Dab1 in Immunohistochemistry applications (Onoune, A., et. al. (2014). Neurosci Res. 88:23-27). |
Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN227625
|
MilliporeSigma
MABN227625UL |
25 μL |
Each for $192.34
|
|
|||||
MABN2276100
|
MilliporeSigma
MABN2276100UL |
100 μL |
Each for $482.80
|
|
|||||
Description
Anti-Dab1, clone 4E12, Cat. No. MABN2276, is a highly specific rat monoclonal antibody that targets Disabled homolog 1 and has been tested in Immunohistochemistry, Immunoprecipitation, and Western Blotting.
Disabled homolog 1 (UniProt: P97318; also known as DAB1) is encoded by the Dab1 gene (Gene ID: 13131) in murine species. Dab 1 is mainly expressed in the brain and serves as an adapter molecule involved in neural development where it is reported to direct the migration of cortical neurons past previously formed neurons to reach their proper layer. It is a cytosolic protein that is shown to be essential component of the reelin signaling pathway. It is shown to bind to the cytoplasmic tail of Reelin receptor, APO E receptor 2, and to LDL receptor. Dab1 can be phosphorylated on Tyr198, Tyr220, and Tyr232 upon reelin induction in embryonic neurons. It can also be phosphorylated on Ser524, but this phosphorylation is independent of reelin signaling. Tyrosine phosphorylation of Dab1 by Src family tyrosine kinases is considered to be the most critical downstream event in Reelin signaling. Dab1 contains a phosphotyrosine interaction domain (PID) (aa 36-189), which specifically binds to the Asn-Pro-Xaa-Tyr(P) motif found in many tyrosine-phosphorylated proteins. Seven different isoforms of Dab1 have been identified that are generated via alternative splicing. Clone 4E12 binds to an epitope within amino acids 161-320. (Ref.: Onoue, A., et al. (2014). Neurosci. Res. 88; 23-27).Specifications
Dab1 | |
Western Blotting Analysis: A representative lot detected Dab1 in Western Blotting applications (Onoune, A., et. al. (2014). Neurosci Res. 88:23-27; Nakamura, K., et. al. (2016). Neuroscience. 336:20-29).Immunoprecipitation Analysis: A 1:500 dilution from a representative lot immunoprecipitated Dab1 in cerebral cortex tissue lysate from P20 mouse (Courtesy of Prof. Mitsuharu Hattori, Nagoya City University).Immunohistochemistry Analysis: A representative lot detected Dab1 in Immunohistochemistry applications (Onoune, A., et. al. (2014). Neurosci Res. 88:23-27). | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
Disabled homolog 1 | |
Dab1 | |
IgG2a κ | |
Protein G purified | |
Clone 4E12 is a rat monoclonal antibody that detects murine Disabled homolog 1 (Dab1). |
4E12 | |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rat | |
Neuroscience | |
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_034144.1 | |
GST-tagged full-length recombinant murine Disabled homolog 1 expressed in E. coli. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title