Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dab1, Rat anti-Mouse, Clone: 4H11, Millipore Sigma™

Catalog No. MABN227725 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN227725 25 μL
MABN2277100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN227725 Supplier MilliporeSigma Supplier No. MABN227725UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Dab1, clone 4H11, Cat. No. MABN2277, is a highly specific rat monoclonal antibody that targets Disabled homolog 1 and has been tested for use in Immunoprecipitation, Immunohistochemistry, and Western Blotting.

Disabled homolog 1 (UniProt: P97318; also known as DAB1) is encoded by the Dab1 gene (Gene ID: 13131) in murine species. Dab 1 is mainly expressed in the brain and serves as an adapter molecule involved in neural development where it is reported to direct the migration of cortical neurons past previously formed neurons to reach their proper layer. It is a cytosolic protein that is shown to be essential component of the reelin signaling pathway. It is shown to bind to the cytoplasmic tail of Reelin receptor, APO E receptor 2, and to LDL receptor. Dab1 can be phosphorylated on Tyr198, Tyr220, and Tyr232 upon reelin induction in embryonic neurons. It can also be phosphorylated on Ser524, but this phosphorylation is independent of reelin signaling. Tyrosine phosphorylation of Dab1 by Src family tyrosine kinases is considered to be the most critical downstream event in Reelin signaling. Dab1 contains a phosphotyrosine interaction domain (PID) (aa 36-189), which specifically binds to the Asn-Pro-Xaa-Tyr(P) motif found in many tyrosine-phosphorylated proteins. Seven different isoforms of Dab1 have been identified that are generated via alternative splicing. Clone 4H1 binds to an epitope within amino acids 281-460. (Ref.: Onoue, A., et al. (2014). Neurosci. Res. 88; 23-27).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dab1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4H11
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A representative lot detected Dab1 in Immunohistochemistry applications (Onoune, A., et. al. (2014). Neurosci Res. 88:23-7).Western Blotting Analysis: A representative lot detected Dab1 in Western Blotting applications (Onoune, A., et. al. (2014). Neurosci Res. 88:23-7; Nakamura, K., et. al. (2016). Neuroscience. 336:20-29).Immunoprecipitation Analysis: A 1:500 dilution from a representative lot immunoprecipitated Dab1 in cerebral cortex tissue lysate of P20 mouse (Courtesy of Prof. Mitsuharu Hattori, Nagoya City University).
Formulation Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Disabled homolog 1
Gene Symbols Dab1
Host Species Rat
Immunogen GST-tagged full-length recombinant murine Disabled homolog 1 expressed in E. coli.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_034144.1
Test Specificity Clone 4H11 is a rat monoclonal antibody that detects murine Disabled homolog 1 (Dab1).
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.