Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102914 Supplier Invitrogen™ Supplier No. PA5102914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DAB2.

DAB2 mRNA is expressed in normal ovarian epithelial cells but is down-regulated or absent from ovarian carcinoma cell lines. The 770-amino acid predicted protein has an overall 83% identity with the mouse p96 protein, a putative mitogen-responsive phosphoprotein; homology is strongest in the amino-terminal end of the protein in a region corresponding to the phosphotyrosine interaction domain. The down-regulation of DAB2 may play an important role in ovarian carcinogenesis. This gene was initially named DOC2 (for Differentially expressed in Ovarian Cancer) and is distinct from the DOC2A and DOC2B genes (for double C2-like domains, alpha and beta).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAB2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DAB2
Gene Accession No. P98078, P98082
Gene Alias 5730435J12Rik; AA960054; Adaptor molecule disabled-2; AI957090; C9; D15Wsu122e; D630005B22Rik; Dab, mitogen-responsive phosphoprotein, homolog 2; Dab2; DAB2, clathrin adaptor protein; differentially expressed in ovarian carcinoma 2; Differentially-expressed protein 2; disabled 2, mitogen-responsive phosphoprotein; disabled homolog 2; disabled homolog 2 mitogen-responsive phosphoprotein; disabled homolog 2, mitogen-responsive phosphoprotein; Doc2; Doc-2; DOC-2 p82 isoform; FLJ26626; Mitogen-responsive phosphoprotein; p96
Gene Symbols DAB2
Host Species Rabbit
Immunogen A synthesized peptide derived from human DAB2(Accession P98082), corresponding to amino acid residues F738-A770.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13132, 1601
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.