Learn More
Invitrogen™ DAB2 Polyclonal Antibody

Description
Predicted to react with rat based on sequence homology.
Specifications
Specifications
| Antigen | DAB2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | DAB2 |
| Gene Accession No. | P98078 |
| Gene Alias | 5730435J12Rik; AA960054; Adaptor molecule disabled-2; AI957090; C9; D15Wsu122e; D630005B22Rik; Dab, mitogen-responsive phosphoprotein, homolog 2; Dab2; DAB2, clathrin adaptor protein; differentially expressed in ovarian carcinoma 2; Differentially-expressed protein 2; disabled 2, mitogen-responsive phosphoprotein; disabled homolog 2; disabled homolog 2 mitogen-responsive phosphoprotein; disabled homolog 2, mitogen-responsive phosphoprotein; Doc2; Doc-2; DOC-2 p82 isoform; FLJ26626; Mitogen-responsive phosphoprotein; p96 |
| Gene Symbols | DAB2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.