Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAB2IP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313027
This item is not returnable.
View return policy
Description
DAB2IP Polyclonal antibody specifically detects DAB2IP in Human samples. It is validated for Western BlotSpecifications
| DAB2IP | |
| Polyclonal | |
| PBS, 1% BSA, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AIP1FLJ39072, ASK-interacting protein 1, DAB2 interacting protein, DAB2 interaction protein, DAB2-interacting protein, DIP1/2, disabled homolog 2-interacting protein, DOC-2/DAB2 interactive protein, KIAA1743AF9Q34ASK1-interacting protein 1, nGAP-like protein | |
| Recombinant protein encompassing a sequence within the C-terminus region of human DAB2IP. The exact sequence is proprietary. | |
| 100 μg | |
| Apoptosis, Zinc Finger | |
| 153090 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction