Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAB2IP Monoclonal Antibody (1626CT702.263.21)
GREENER_CHOICE

Catalog No. PIMA537700
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537700 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537700 Supplier Invitrogen™ Supplier No. MA537700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DAB2IP Monoclonal Antibody for Western Blot

DAB2IP is a Ras GTPase-activating protein (GAP) that acts as a tumor suppressor. The DAB2IP gene is inactivated by methylation in prostate and breast cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAB2IP
Applications Western Blot
Classification Monoclonal
Clone 1626CT702.263.21
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Dab2ip
Gene Accession No. Q3UHC7, Q5VWQ8
Gene Alias 2310011D08Rik; AF9Q34; AI480459; AIP1; AIP-1; ASK1-interacting protein 1; ASK-interacting protein 1; DAB2 interacting protein; DAB2 interaction protein; DAB2-interacting protein; Dab2ip; DIP1/2; disabled 2 interacting protein; disabled 2 interacting protein long form; disabled homolog 2 interacting protein; disabled homolog 2-interacting protein; DOC2/DAB2 interactive protein; DOC-2/DAB2 interactive protein; DOC-2/DAB-2 interactive protein; Kiaa1743; mKIAA1743; nGAP-like protein
Gene Symbols Dab2ip
Host Species Mouse
Immunogen recombinant protein between 782-1038 amino acids from human DAB2IP.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153090, 69601
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.